Learn More
Thermo Scientific™ Lambda DNA/EcoRI plus HindIII Marker, ready-to-use
Description
Lambda DNA/EcoRI+HindIII Marker, ready-to-use is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
Lambda DNA/EcoRI+HindIII Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading (after heating) onto agarose gels.
Highlights
- Sizing and approximate quantification of large DNA fragments
- Sharp bands
- Premixed with 6X DNA Loading Dye for a direct loading (after heating) and for room temperature storage
- Supplied with loading dye for sample DNA
Specifications
Specifications
| Concentration | 6X |
| Gel Type | Agarose |
| Ready to Load | Yes |
| Gel Compatibility | Agarose Gels |
| Quantity | 5 x 50 μg |
| Product Type | DNA/ECORI Plus Hind III Marker |
Frequently Asked Questions (FAQs)
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.