Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RNase A/T1 Mix
GSA_VA

Catalog No. FEREN0551 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0551 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREN0551 Supplier Thermo Scientific™ Supplier No. EN0551
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Get the RNA degradation activity of both RNase A and RNase T1 with Thermo Scientific™ RNase A/T1 Mix.

Thermo Scientific RNase A/T1 Mix combines the RNA degradation activity of both RNase A and RNase T1. The RNase A specifically hydrolyzes RNA at C and U residues; RNase T1 specifically hydrolyzes RNA at G residues (see Figure 1 in Supporting Data [see Reference 1]).

Highlights

  • Higher level of RNA degradation than with RNase A and RNase T1 separately
  • Free of DNase activity - It is not necessary to heat the mix before use

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Enzyme RNase
Quantity 1 mL
Product Type RNase A/T1 Mix
What is your recommended RNase digestion mixture with Thermo Scientific RNase A/T1 Mix for RNase protection assay?

We recommend 10 mM Tris-HCl (pH 7.5), 300 mM NaCl, 5 mM EDTA (pH 7.5), 20 µL of RNase A/T1 Mix per 1 mL of reaction mixture.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.