Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RNase I (10 U/μL)
GSA_VA

Catalog No. p-4521367
Change view
Click to view available options
Quantity:
1,000 units
5,000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0601 1,000 units
FEREN0602 5,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREN0601 Supplier Thermo Scientific™ Supplier No. EN0601
Only null left
Add to Cart
Edge
Add to Cart

Ribonuclease I (RNase I) is an endoribonuclease that hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates.

Ribonuclease I (RNase I), an endoribonuclease, preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

  • Stable and active under a wide variety of reaction conditions
  • Can be heat inactivated in 30 minutes at 100°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Enzyme RNase
Quantity 1,000 units
Product Type RNase I
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.