Learn More
Thermo Scientific™ T7 DNA Polymerase (10 U/μL)
Catalog No. FEREP0081 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
300 units
Catalog No. Quantity
FEREP0081 300 units
1 options
Catalog No. FEREP0081 Supplier Thermo Scientific™ Supplier No. EP0081
Add to Cart
Add to Cart
Catalyze DNA synthesis in the 5'≥3' direction with T7 DNA Polymerase, a template dependent DNA polymerase.
T7 DNA Polymerase, a template dependent DNA polymerase, catalyzes DNA synthesis in the 5'≥3' direction. It is a highly processive DNA polymerase allowing continuous synthesis of long stretches of DNA. The enzyme also exhibits a high 3'≥5' exonuclease activity towards single- and double-stranded DNA.
Highlights
  • Strong 3'=>5' exonuclease activity, approximately 1000-fold greater than Klenow Fragment (see Reference 1)
  • Active in restriction enzyme buffers
TRUSTED_SUSTAINABILITY
Specifications
Polymerase T7 DNA Polymerase
Quantity 300 units
For Research Use Only. Not for use in diagnostic procedures.