Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Van91I (PflMI) (10 U/μL)

Catalog No. p-4522197
Change view
Click to view available options
Quantity:
2,000 units
400 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERER0711 400 units
FERER0712 2,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERER0711 Supplier Thermo Scientific™ Supplier No. ER0711
Only null left
Add to Cart
Edge
Add to Cart

Cut at CCANNNN^NTGG sites with Van91I (PflMI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: AccB7I, BasI, PflMI).

Lambda DNA digested with Van91I (PflMI), 0.7% agarose, 14 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..C C A NN N N^N T G G...3'

3'..G G T N^N N NN A C C...5'

Conditions for 100% Activity

  • 1X Buffer R: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/ml BSA
  • Incubate at 37°C

Storage Buffer

  • Van91I is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with Van91I, more than 90% of DNA fragments can be ligated and recut

Methylation Effects

  • Dam: never overlaps – no effect
  • Dcm: may overlap – blocked
  • CpG: never overlaps – no effect
  • EcoKI: never overlaps – no effect
  • EcoBI: never overlaps – no effect&

Digestion of Agarose-embedded DNA

  • Minimum 5 units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

Van91I is blocked by overlappingdcm methylation. To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Van91I (PflMI)
Compatible Buffer 10X Buffer R
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 400 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.