Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ X-Gluc

Catalog No. FERR0852
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0852 0.5 g
FERR0851 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR0852 Supplier Thermo Scientific™ Supplier No. R0852
Add to Cart
Add to Cart

Effectively screen colonies with X-Gluc, a substrate for ß-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene.

X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).

Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).

Recommended for:

  • Detection of GUS expression in plant cells and tissues (1)
  • Detection of infections caused by E.coli
  • Detection of bacterial contamination in food and water samples

Note:

Preparation of a 0.1M stock solution in dimethylformamide or dimethylsulfoxide is recommended.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Product Type X-Gluc
Quantity 0.5 g
Substrate Properties Chemical Substrate
Substrate Type Beta-Glucuronidase (GUS) Substrate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.