Learn More
Ferno™ Fastrap Quick Restraint System
SDP
Catalog No. 191305703
Encompass
Change view
Click to view available options
Product Type:
Restraint System
Catalog No. Product Type
19-130-5703 Restraint System
1 options
Catalog No. 19-130-5703 Supplier Ferno™ Supplier No. 0313916
Specifications
Product Type Restraint System
For Use With (Equipment) Najo™ backboard and scoop stretcher
Includes 21 in. Wrist strap, 66 in. chest, pelvic, thigh straps, 46 in. foot strap, 60 in. V-strap
Weight (English) 2 lb.
Weight (Metric) 0.69 kg