Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferno™ Fastrap™ Quick Restraint System for Backboards
SDP

Catalog No. 19811421
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Restraint System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-811-421 Restraint System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-811-421 Supplier Ferno™ Supplier No. 770
May include imposed supplier surcharges.
Add to Cart
Add to Cart

To secure patients to backboards

  • Ten-point, six-strap
  • Color-coded for fast and easy application
  • Hook and loop nylon closures
  • Sdjustable size from pediatric to adult
  • Compatible with the Millennia Backboard, NAJO Backboard series, Wooden Backboard Series, 65 Scoop, and 65 EXL Scoop Stretchers

Specifications

Product Type Restraint System
For Use With (Equipment) Najo™ backboard and scoop stretcher
Includes 21 in. Wrist strap, 66 in. chest, pelvic, thigh straps, 46 in. foot strap, 60 in. V-strap
Weight (English) 2 lb.
Weight (Metric) 0.69 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.