Learn More
Kingfisher Biotech Inc Ferret/Mink LIF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2133FT100
Encompass_Preferred
The Ferret/Mink Leukemia Inhibitory Factor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ferret/Mink Leukemia Inhibitory Factor applications are for cell culture, ELISA standard, and Western Blot Control. Ferret/Mink Leukemia Inhibitory Factor yeast-derived recombinant protein can be purchased in multiple sizes. Ferret/Mink Leukemia Inhibitory Factor Specifications: (Molecular Weight: 19.7 kDa) (Amino Acid Sequence: TPLPITPVNA TCATRHPCHS NLMNQIRNQL AQLNGSANAL FILYYTAQGE PFPNNLDKLC GPNVTDFPPF HANGTEKTRL VELYRIIAYL GASLGNITRD QKVLNPNALS LHSKLNATAD ILRGLLSNVL CRLCNKYHVA HVDVAYGPDT SGKDVFQKKK LGCQLLGKYK QVIAVVAQAF (180)) (Gene ID: 101674557). For research use only. Made in the USA
Catalog No. 50-253-2378
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.