Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, MP Biomedicals

Catalog No. ICN15349991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15349991 1 kg
ICN15349980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15349991 Supplier MP Biomedicals, Inc Supplier No. 0215349991/DEL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Chlorure Perrique, Iron (III) Chloride
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Black
Physical Form Powder
Quantity 1 kg
Formula Weight 162.21
Density 2.898g/cm3 at 25°C
Chemical Name or Material Ferric chloride, anhydrous, Chemistry
EINECSNumber 231-096-4
storageNote1 Store desiccated. Store under nitrogen
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.