Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Citrate, Purified, Spectrum™ Chemical
SDP

Catalog No. 18602669 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-669 2.5 kg Poly Pail
18-602-667 125 g Poly Bottle
18-602-668 500 g Poly Bottle
18-602-670 12 kg Poly Pail
18-602-671 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-669 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F10282.5KG
Only null left
Add to Cart
Add to Cart

F1028, 2338-05-8, Approx. FeC6H5O7·xH2O

Spectrum™ Chemical Ferric Citrate, Purified is a powder with a green to reddish-brown appearance. It can also be used in water purification, printing and an acidity regulator.

Chemical Identifiers

CAS 2338-05-8
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Grade Purified
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.