Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Solution, 5% w/v, Ricca Chemical Company
SDP

Catalog No. R31370004B Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R31370004B 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R31370004B Supplier Ricca Chemical Company Supplier No. R31370004B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10028-22-5 , 7664-93-9 , 7732-18-5

Specifications

Color Yellow
Boiling Point ~100°C
Concentration 5% w/v
Quantity 4 L
Sterility Non-sterile
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Ferric Sulfate Solution
ShelfLife 18 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.