Learn More
Description
Ferric Subsulfate USP is a reddish brown liquid, odorless or nearly so. It is affected by light.
Alternative name(s): ferric subsulfate solution
Specifications
Specifications
| Chemical Name or Material | Ferric Subsulfate USP (Monsels Solution) |
| Packaging | Bottle, Amber Glass, Boston Round, 500mL, 38-439 |
| CAS | 1310-45-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 737.704 |
| Quantity | 500 mL |
| Percent Purity | 20 to 22 g of iron per 100 mL |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
