Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferriprotoporphyrin IX chloride, Thermo Scientific™

Catalog No. AA3206103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3206103 1 g
AA3206106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3206103 Supplier Thermo Scientific Chemicals Supplier No. 3206103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15489-47-1
MDL Number MFCD00010726
Synonym protoporphyrin 2-, 7,12-diethenyl-3,8,13,17-tetramethylporphyrin-2,18-dipropanoate, 2,7,13,17-tetramethyl-3,18-diethenyl 21h,23h-porphyrin-8,12-bispropionate, 3,8-diethenyl-2,7,12,18-tetramethyl-21h,23h-porphyrin-13,17-dipropanoate, 3-7,12-bis ethenyl-3,8,13,17-tetramethyl-18-3-oxido-3-oxopropyl-22,23-dihydroporphyrin-2-yl propanoate

Specifications

Quantity 1 g
Formula Weight 651.93
Physical Form Crystalline
Chemical Name or Material Ferriprotoporphyrin IX chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.