Learn More
Apexbio Technology LLC ferritin heavy chain fragment [Multiple species] 10mg
SDP
Supplier:  Apexbio Technology LLC A106910
Encompass_Preferred
Ferritin heavy chain fragment is a protein subunit fragment associated with iron storage and metabolism It is derived from H-ferritin a component predominantly expressed in heart kidney placenta and various tumor cells and forms part of the larger 24-subunit ferritin complex The ferritin heavy chain exhibits ferroxidase activity catalyzing the oxidation of Fe to Fe thereby sequestering intracellular iron and reducing the pool of catalytically active free iron Based on its biochemical attributes ferritin heavy chain fragment is widely utilized as a research tool or biomarker in studies investigating iron metabolism disorders iron overload diseases tumor biology inflammation and immune modulation in oncology
Catalog No. 50-000-95129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.