Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Ferritin ELISA Kit

Catalog No. EHFTLX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHFTLX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHFTLX10 Supplier Invitrogen™ Supplier No. EHFTLX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. Ferritin is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in the light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. The ferritin gene has multiple pseudogenes, and ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. Ferritin is a holoenzyme shell with a molecular weight of about 450 kDa, and consists of 24 subunits of two types, H(heavy) and L(light), and capable of storing up to 4,500 atoms of ferric iron.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02792, P02794
Assay Range 0.81 to 200 ng/mL
Assay Sensitivity 600 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2495, 2512
Gene Symbol FTH1, FTL
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target Ferritin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.