Learn More
Description
MA5-14733 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MA5-14733 Immunogen is human liver ferritin. MA5-14733 detects ferritin which has a predicted molecular weight of approximately 19kDa. Product MA514733 is a smaller package size of MIF2501 (formerly sold as a Seradyn product).
Specifications
Specifications
| Antigen | Ferritin |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 101 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | FTL |
| Gene Accession No. | P02792, P02794 |
| Gene Alias | apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3; Ferritin subunit H; ferritin, heavy polypeptide 1; ferritin, light polypeptide; FHC; frih; fril; FTH; Fth1; FTHL6; FTL; HFE5; H-ferritin; HFt; L apoferritin; LFTD; MFH; MGC71996; NBIA3; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
