Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AA3925404
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3925404 2 g
AA3925409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3925404 Supplier Thermo Scientific Chemicals Supplier No. 03925404
Add to Cart

missing translation for 'validMainframeMsg'

Ferrocenecarboxaldehyde, is used to prepare chiral ferrocene aziridinylmethanols for selective azomethine ylide cycloaddtion. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12093-10-6
Molecular Formula C11H10FeO
Molecular Weight (g/mol) 214.05
MDL Number MFCD00001429
InChI Key UQTCQJVPLIVCAX-UHFFFAOYNA-N
Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
IUPAC Name (cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
SMILES [Fe].c1cccc1.O=Cc1cccc1

Specifications

Melting Point 124°C
Quantity 2 g
Sensitivity Air sensitive
Solubility Information It is soluble in water.
Formula Weight 214.05
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Ferrocenecarboxaldehyde
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 235-158-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.