Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals

Catalog No. AAA1687403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1687403 1 g
AAA1687406 5 g
AAA1687414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1687403 Supplier Thermo Scientific Chemicals Supplier No. A1687403
Add to Cart

Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-42-7
Molecular Formula C11H10FeO2
Molecular Weight (g/mol) 230.04
MDL Number MFCD00001430
InChI Key OUPWBVGTMCICLR-UHFFFAOYSA-N
Synonym Carboxyferrocene; (Carboxycyclopentadienyl)cyclopentadienyliron
IUPAC Name Ferrocenecarboxylic acid
SMILES [Fe].c1cccc1.OC(=O)c1cccc1

Specifications

Melting Point ∼210°C (decomposition)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 230.05
Percent Purity 98%
Chemical Name or Material Ferrocenecarboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-040-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.