Learn More
TARGETMOL CHEMICALS INC FERROHEME 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T894810MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Ferroheme (protoheme) is the color-furnishing portion of hemoglobin. It is found free in tissues and as the prosthetic group in many hemeproteins. A Ferroheme or haem is a prosthetic group that consists of an iron atom contained in the center of a large heterocyclic organic ring called a porphyrin. purity: 100%
Catalog No. 50-257-6216
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.