Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroin Indicator Solution, 0.025M, Honeywell Fluka™

Catalog No. 60046933 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-933 250 mL Glass Bottle
60-046-934 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-933 Supplier Honeywell Chemicals Supplier No. 46270250ML
Only null left
Add to Cart
Add to Cart

Redox Indicator, 0.025 M

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
PubChem CID 84567
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 0°C
Boiling Point 100°C
Linear Formula [Fe(C12H8N2)3]SO4
UN Number NONH for all modes of transport
Formula Weight 692.52g/mol
Odor Odorless
Packaging Glass Bottle
Density 1.006g/mL at 20°C
Color Red
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Ferroin indicator solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.