Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroin Indicator Solution, 0.025M Orthophenanthroline TS, Reagents

Catalog No. 01336757
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-757 500 mL
01-336-759 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-757 Supplier Reagents Holdings Llc Supplier No. C6110900500C
Add to Cart
Add to Cart

Ferroin Indicator Solution, 0.025M Orthophenanthroline TS, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ferroin Indicator Solution, 0.025M Orthophenanthroline TS
CAS 7732-18-5, 5144-89-8, 5144-89-8
Boiling Point 100°C
Synonym O-Phenanthroline, 1,10-Phenanthroline
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.01
Color Dark Red
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.