Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ferroportin Monoclonal Antibody (C2), Invitrogen™

Catalog No. p-7240831
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543825 20 μL
PIMA543824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543825 Supplier Invitrogen™ Supplier No. MA543825
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ferroportin
Applications Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SLC40A1
Gene Accession No. Q9NP59
Gene Alias CAR1; Cell adhesion regulator; duodenal-specific; Dusg; Ferroportin; ferroportin 1; ferroportin-1; Fpn1; Fpt1; HFE4; IREG1; iron regulated gene 1; iron-regulated transporter 1; Metal transporter protein 1; metal transporting protein 1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; polycythaemia; Slc11a3; SLC11A3 iron transporter; Slc39a1; Slc40a1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; solute carrier family 40 member 1
Gene Symbols SLC40A1
Host Species Mouse
Immunogen Trp127-Val321 (Accession Q9NP59), with N-terminal His and GST Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.