Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ferroportin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564232 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564232 Supplier Invitrogen™ Supplier No. PA564232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKAGLKEEET ELKQLNLHKD TEPKPLEGTH LMGVKDSNIH ELEHEQEPTC ASQMAEPFRT FRDGWVSYYN Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ferroportin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC40A1
Gene Accession No. Q9NP59
Gene Alias CAR1; Cell adhesion regulator; duodenal-specific; Dusg; Ferroportin; ferroportin 1; ferroportin-1; Fpn1; Fpt1; HFE4; IREG1; iron regulated gene 1; iron-regulated transporter 1; Metal transporter protein 1; metal transporting protein 1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; polycythaemia; Slc11a3; SLC11A3 iron transporter; Slc39a1; Slc40a1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; solute carrier family 40 member 1
Gene Symbols SLC40A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC40A1. Recombinant protein control fragment (Product #RP-104220).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.