Learn More
Specifications
Specifications
| Gene ID (Entrez) | 30061 |
| Species | Human |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-21502 |
| Gene Alias | Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 |
| Gene Symbol | SLC40A1 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 62.5 kDa |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
