Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate Hexahydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCF0300500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCF0300500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCF0300500 Supplier Ricca Chemical Company Supplier No. RDCF0300500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Blue-Green
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Ferrous Ammonium Sulfate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.