Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate Hexahydrate, ACS Reagent, Reagents

Catalog No. 01337969
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-969 100 g
01-337-968 12 kg
01-337-966 3 kg
01-337-965 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-969 Supplier Reagents Holdings Llc Supplier No. C1206000100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous Ammonium Sulfate Hexahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Iron(II) Ammonium Sulfate Hexahydrate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Boiling Point 100°C to 110°C
Melting Point 100°C to 110°C
Color Pale Blue-Green
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.864
Chemical Name or Material Ferrous Ammonium Sulfate Hexahydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.