Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate, Reagents

Catalog No. 01335291
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-291 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-291 Supplier Reagents Holdings Llc Supplier No. CF002800500C
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous Ammonium Sulfate, 0.00282 N 1 mL = 0.1 mg Cl2, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7783-85-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Iron(II) Ammonium Sulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Light Green
pH <7
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.001
Chemical Name or Material Ferrous Ammonium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.