Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Chloride Tetrahydrate, Reagent (Iron (II) Chloride Tetrahydrate), Reagents

Catalog No. 01337962
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-962 500 g
01-337-964 100 g
01-337-963 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-962 Supplier Reagents Holdings Llc Supplier No. C1206500500C2
Add to Cart
Add to Cart

Ferrous Chloride Tetrahydrate, Reagent (Iron (II) Chloride Tetrahydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym Iron(II) Chloride Tetrahydrate
IUPAC Name tetrahydroxyironbis(ylium) dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Fe++]

Specifications

Boiling Point Sublimes
Melting Point 670°C to 674°C
Color Green to Blue-Green
Physical Form Solid
Quantity 500 g
Percent Purity ≥ 98%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.93
Chemical Name or Material Ferrous Chloride Tetrahydrate, Reagent (Iron (II) Chloride Tetrahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.