Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferrous Sulfate Heptahydrate
SCI_ED

Catalog No. S25325 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25325 100 g Solid
S25325A 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25325

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Color Blue-Green
Quantity 100 g
Assay Percent Range >77%
Linear Formula FeSO4·7H2O
Formula Weight 278.01
Packaging Solid
Density 1.898g/mL
Chemical Name or Material Ferrous Sulfate, Heptahydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.