Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18602713 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-713 500 g Amber Glass Bottle
18-602-711 125 g Amber Glass Bottle
18-602-712 6 x 125 g Amber Glass Bottle
18-602-715 2.5 kg Amber Glass Bottle
18-602-714 6 x 500 g Amber Glass Bottle
18-602-716 4 x 2.5 kg Amber Glass Bottle
18-602-717 12 kg Poly Pail
18-602-718 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-602-713 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1060500GM
Add to Cart
Add to Cart

F1060, 7782-63-0, FeSO4·7H2O

Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.