Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate Solution 40% (w/v), for AOAC Method 932.05 for Citric Acid in Milk, Gravimetric Method, Fisher Chemical
SDP

Catalog No. SF9100500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF9100-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF9100-500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7782-63-0 , 7732-18-5 , 7664-93-9
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name λ2-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Min % 27.65, 69.0, 1.27
CAS Max % 28.78, 72.0, 1.32
Percent Purity 40%
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material Ferrous Sulfate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.