Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Ferrules HP style graphite, I.D. 0.8 mm

Catalog No. 111011546 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
For 0.53 mm I.D. Columns in HP Instruments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
11-101-1546 For 0.53 mm I.D. Columns in HP Instruments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-1546 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2355471PAK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.