Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferulic Acid, 99.4%, MP Biomedicals™

Catalog No. ICN10168505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10168505 5 g
ICN10168525 25 g
ICN10168580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10168505 Supplier MP Biomedicals, Inc Supplier No. 0210168505
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1135-24-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
InChI Key KSEBMYQBYZTDHS-HWKANZROSA-N
Synonym ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
PubChem CID 445858
ChEBI CHEBI:17620
IUPAC Name (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=CC(=C1)C=CC(=O)O)O

Specifications

Assay Percent Range 99.4%
Quantity 5 g
Formula Weight 194.19
Physical Form Crystalline
Chemical Name or Material Ferulic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.