Learn More
AdipoGen Ferutinin high purity
SDP
Supplier:  AdipoGen AGCN20007M001
Encompass_Preferred
Chemical. CAS 41743-44-6. Formula C22H30O4. MW 358.5. Semisynthetic. Potent, naturally occuring non-steroid estrogenic compound. Tool to promote mitochondrial calcium overload and to promote calcium-dependent opening of the mitochondrial permeability transition pore mPTP. Strong agonist for estrogen receptor ERalpha and agonist/antagonist for ERbeta. Calcium ionophoretic Apoptosis modulator. Antiproliferative. Increases nitric oxide synthase activity and phosphoinositides breakdown in nervous tissue. Shows aphrodisiac and anti-sexual impotence activities. Anti-osteoporotic. Prevents osteoporosis caused by severe estrogen deficiency. Modest AChE inhibitor.
Catalog No. 50-256-2700
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.