Learn More
Apexbio Technology LLC Fesoterodine Fumarate 286930-03-8 50mg
SDP
Supplier:  Apexbio Technology LLC B161850
Encompass_Preferred
Fesoterodine Fumarate (CAS 286930-03-8) is a synthetic antimuscarinic agent that acts as a competitive antagonist at muscarinic acetylcholine receptors particularly the M3 subtype in the bladder detrusor muscle By inhibiting these receptors the compound modulates involuntary bladder contractions reducing symptoms associated with overactive bladder syndrome such as urgency and frequency Fesoterodine Fumarate is utilized in biomedical research to study cholinergic signaling mechanisms involved in urinary function and to explore pharmacological interventions targeting muscarinic pathways in urological disorders
Catalog No. 50-000-92709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.