Learn More
Perkin Elmer US LLC Festoon Lamp - 3 W
Supplier:  Perkin Elmer US LLC 04969486
Encompass_Preferred
Our Fluorescence Spectrometers use Pulsed Xenon Discharge lamps. These reliable, stable lamps provide remarkable sensitivity and can achieve a peak intensity exceeding 1 KW. To maintain the original stability and sensitivity of the instrument only use PerkinElmer replacment lamps. The quartz envelops of these lamps are pressurized and require extreme caution. Replacement by PerkinElmer Service Engineers is recommended.
Catalog No. 50-211-3500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.