Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Fetal Bovine Serum, Premium, New Zealand Origin
GREENER_CHOICE

Catalog No. MT35098CV Shop All Corning Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
Serum Treatment:
Gamma-irradiated
Heat-inactivated
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Serum Treatment
MT35098CV 500 mL Gamma-irradiated
MT35088CF 50 mL Heat-inactivated
MT35078CF 50 mL None
MT35088CV 500 mL Heat-inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. MT35098CV Supplier Corning™ Supplier No. 35098CV
Add to Cart
Add to Cart
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Collected at government registered facilities in New Zealand

  • Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
  • True pooled and is triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
  • Validated process used to heat inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -40°C to -10°C
Species Bovine
Country of Origin New Zealand
Form Liquid
Quantity 500 mL
Serum Treatment Gamma-irradiated
Sterility Sterile
Source New Zealand Origin
Heat Inactivated No
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.