Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Fetal Bovine Serum, Premium, United States Origin
GREENER_CHOICE

Catalog No. MT35073CI Shop All Corning Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Serum Treatment:
Heat-inactivated
None
Tetracycline Negative
Ultra-low IgG
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Serum Treatment
MT35073CI 100 mL Ultra-low IgG
MT35016CF 50 mL Heat-inactivated
MT35015CF 50 mL None
MT35075CF 50 mL Tetracycline Negative
MT35017CV 500 mL Heat-inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MT35073CI Supplier Corning™ Supplier No. 35073CI
Add to Cart
Add to Cart

Collected at government approved abattoirs in the United States

  • True pooled and triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).
  • Validated process used to heath inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
  • Validated process used to gamma irradiate serum within one of two separate dose ranges: 25-40 kGy and 30-45 kGy.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -40°C to -10°C
Species Bovine
Country of Origin United States
Form Liquid
Quantity 100 mL
Serum Treatment Ultra-low IgG
Source United States Origin
Heat Inactivated No
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.