Learn More
Description
- Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
- True pooled and triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
Specifications
Specifications
| Content And Storage | -40°C to -10°C |
| Species | Bovine |
| Country of Origin | United States |
| Form | Liquid |
| Quantity | 50 mL |
| Serum Treatment | None |
| Sterility | Sterile |
| Source | USDA Origin |
| Heat Inactivated | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
