Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Fetal Bovine Serum (U.S.), Super Low IgG

Catalog No. SH308980340 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Irradiated, 25 to 40 doses
None
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH308980340 500 mL Irradiated, 25 to 40 doses
SH3089802 100 mL None
SH3089803 500 mL None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH308980340 Supplier Cytiva Supplier No. SH30898.03IR2540
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production

Fetal bovine serum (FBS) has natural levels of IgG that are too high for some cell culture applications. Lots can be prescreened to find the lowest naturally occuring levels, but as a biological product, FBS is frequently in short supply. Even lots with naturally low IgG levels may still contain too much for some applications. Examples of typical IgG levels in bovine serum products can be found in the table below.

  • Performance is equivalent to non-processed FBS
  • Suitable for applications where low levels of specific antibody are required (e.g. propagation of bovine viruses, monoclonal antibody production)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C
Quantity 500 mL
Processing Irradiated, 25 to 40 doses
Heat Inactivated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.