Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fetal Hemoglobin mAb Test Kit, R-PE Conjugate

Catalog No. 501137426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7426 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-113-7426 Supplier Invitrogen™ Supplier No. HFH04
  • $860.00
    $644.65 / Each of 1

    Save $215.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Fetal Hemoglobin (HbF) Test is based on a monoclonal antibody directed to fetal hemoglobin (hemoglobin F).

The Fetal Hemoglobin (HbF) Test is based on a monoclonal antibody directed to fetal hemoglobin (hemoglobin F). This antibody is conjugated with R-phycoerythrin (R-PE). R-PE (phycoerythrin) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence emission without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients. Use of R-PE in antibody conjugates results in probes with greatly enhanced detectability.

This reagent is intended for the identification and subsequent enumeration of fetal red blood cells. Fetal cells are identified by the presence of fetal hemoglobin by a flow cytometric method. The Fetal Hemoglobin Test may be used as an aid in identifying fetal-maternal hemorrhage and determining the need for immunoprophylaxis with immune globulin.

Antibody Specifications

  • Description: Fetal Hemoglobin F (HbF), mouse IgG1 monoclonal
  • Label: R-PE (phycoerythrin)
  • Validated Application: Flow Cytometry
  • Species Reactivity: Human
  • Ex/Em and laser configuration: 496/578 nm, blue laser
  • Clone ID: HbF-1

Clinical Relevance
The Fetal Hemoglobin Test contains a monoclonal antibody directed to HbF that specifically recognizes fetal hemoglobin found in fetal erythrocytes. The test permits the enumeration of fetal erythrocytes in the maternal circulation as a result of a fetal-maternal hemorrhage, or leakage, of fetal cells into the maternal circulation. Hemorrhage of cells from an Rh+ fetus into the circulation of an Rh- mother may result in the formation of Rh-reactive antibodies in the mother. Rh hemolytic anemia in a subsequent Rh+ fetus may result from placental transfer of antibodies formed in the mother to the fetus. Although the volume of fetal erythrocytes found in the maternal circulation during pregnancy and immediately post-partum is reported to be small and of uncertain clinical significance in many cases, substantial hemorrhage may result from a number of causes including fetal or maternal trauma and placental defects.

Erythrocytes containing fetal hemoglobin may be found in individuals of any age, but with lower amounts of fetal hemoglobin compared to fetal red cells. These cells have been termed F cells. High levels of F cells may also exist in adults with a heterogeneous group of genetic disorders of uncertain etiology referred to as Hereditary Persistence of Fetal Hemoglobin. Other clinical conditions causing significant levels of anemia may also result in elevated levels of F cells. Several clinical conditions have been described that involve increased levels of F cells. These conditions include hereditary anemic diseases such as sickle cell anemia and thalassemia major.

Additional Information
Use the Spectraviewer to help you plan your experiments. Additional general information about Life Technologies flow cytometry products and support is also available.

TRUSTED_SUSTAINABILITY

Specifications

Antibody Conjugate PE and APC, R-PE
Buffer PBS (Phosphate-Buffered Saline), 0.1% BSA
Content And Storage Store in refrigerator (2°C to 8°C).
CE Marker Yes
Detection Method Fluorescence
Format Tube
Type Hemoglobin Test Kit
For Use With (Application) Flow Cytometry Antibodies and Secondary Detection
Label or Dye R-PE
Quantity 100 Tests
Shipping Condition Wet Ice
Species Mouse
Target Gene Fetal Hemoglobin
Validated Application Flow Cytometry
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.