Learn More
Invitrogen™ Fetal Hemoglobin Monoclonal Antibody (HBF-1), FITC, Invitrogen™

Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility. This Fetal Hemoglobin monoclonal antibody specifically recognizes fetal hemoglobin found in fetal erythrocytes.
Specifications
Specifications
| Antigen | Fetal Hemoglobin, Fetal Hemoglobin (HbF) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HBF-1 |
| Conjugate | FITC |
| Formulation | PBS with <10% sucrose, BSA and 0.1% sodium azide |
| Gene | HBG1 |
| Gene Accession No. | P69891 |
| Gene Alias | A-gamma globin; alpha-1 globin; gamma A hemoglobin; gamma globin; gamma-1-globin; Hb F Agamma; HBG1; HBGA; HBGR; HBG-T2; hemoglobin alpha 1; Hemoglobin gamma-1 chain; hemoglobin gamma-a chain; hemoglobin subunit gamma 1; hemoglobin subunit gamma-1; hemoglobin, gamma A; hemoglobin, gamma, regulator of; HSGGL1; PRO2979 |
| Gene Symbols | HBG1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.