Learn More
Medchemexpress LLC Fetal-tau/0N3R Huma 50ug
SDP
Supplier:  Medchemexpress LLC HYP7361850UG
Encompass_Preferred
Tau/0N3R protein is encoded by the Tau/0N3R gene and undergoes regulated alternative splicing to produce different mRNA species Its expression varies in the nervous system according to the maturation and type of neurons Tau/0N3R Protein Human (His solution) is the recombinant human-derived Tau/0N3R protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-80836