Learn More
Description
Sheep anti Bovine asialofetuin antibody recognizes bovine Alpha-2-HS-glycoprotein, also known as asialofetuin or fetuin-A. In a simple antigen down ELISA assay reactivity includes: asialofetuin (1:8000) 100%, fetuin (1:2000) 100%, fetuin (1:8000) 50%.
Specifications
Specifications
| Antigen | Fetuin A |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | AHSG |
| Gene Accession No. | P12763 |
| Gene Alias | 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743 |
| Gene Symbols | AHSG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
