Learn More
Sigma Aldrich Fine Chemicals Biosciences fetuin from fetal bovine s5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F33855G
Encompass
fetuin from fetal bovine serum is a plasma glycoprotein. It is a 64 kDa glycoprotein which is produced by the liver and adipose tissue.A glycoprotein derived from fBS that is used as fetuin has been used at a concentration of 500 mg/ml to supplement serum free f12 medium (along with insulin transferrin and 2-mercaptoethanol) in culture of embryonal carcinoma cells.
Catalog No. 50-291-4182