Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fetuin, From Fetal Bovine Serum

Catalog No. MP21048741 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21048741 100 mg
MP21048742 250 mg
ICN10487401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP21048741 Supplier MP Biomedicals™ Supplier No. 02104874.1
Add to Cart
Add to Cart

Fetuin is a protein containing 12 Cys residues, all involved in disulfide bridges.

  • Formulation: Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%

Applications:

Fetuin bovine is a Protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Type 2 Diabetes Mellitus (T2DM) Studies, Cell Attachment and Proliferation
Formulation Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%
Name Fetuin Protein
Purification Method Ammonium Sulfate Fractionation
Quantity 100 mg
Source Fetal Bovine Serum
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.