Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFA Solution, Toluene Free, Reagents
FFA Solution, Toluene Free, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CF13350020F
Specifications
| FFA Solution, Toluene Free | |
| 67-63-0, 7732-18-5 | |
| Free Fatty Acid Isopropanol | |
| Cubitainer | |
| 20 L | |
| Liquid |
| Colorless to Pink | |
| 83°C | |
| Reagent | |
| 0.781 | |
| Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction