Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FFAR3/GPR41, Mouse, Unlabeled, Clone: 16F4.1,

Catalog No. MABN898MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN898MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN898MI Supplier MilliporeSigma™ Supplier No. MABN898
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Free fatty acid receptor 3 (UniProt O14843; also known as G-protein coupled receptor 41) is encoded by the FFAR3 (also known as GPR41) gene (Gene ID 2865) in human. Free fatty acids (FFAs) act as both energy sources and signaling molecules via G-protein-coupled receptors (GPCRs) called FFARs (FFA receptors). FFAs are categorized by the length of their carbon chains into short-chain FAs (SCFAs; less than 6 carbons), medium-chain FAs (MCFAs; 6–12 carbons), and long-chain FAs (LCFAs; more than 12 carbons). MCFAs and LCFAs activate FFAR1 and FFAR4, whereas SCFAs activate FFAR3 and FFAR2. FFAR3 is an SCFA receptor that can be activated by propionate (C3), butyrate (C4), and valerate (C5). FFAR3 mediates PYY and GLP-1 secretion from endocrine and sympathetic outflow-dependent heart rate regulation and energy expenditure, while β-Hydroxybutyrate, a ketone body produced in the liver during starvation, decreases sympathetic outflow by antagonizing FFAR3. FFAR3 is shown to couple Gαi/o pathway signaling upon activation by propionate binding in primary murine pancreatic beta cells, resulting in an inhibition of glucose-dependent insulin secretion. FFAR3 is a 7-transmembrane (a.a. 20-40, 48-68, 89-111, 133-153, 179-199, 223-243, 259-279) GPCR, having a short N-terminal extracellular portion (a.a. 1-19) and a long C-terminal cytomplasmic tail (a.a. 280-346).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FFAR3/GPR41
Applications Western Blot
Classification Monoclonal
Clone 16F4.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene FFAR3, GPR41
Gene Accession No. O14843
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal cytoplasmic domain sequence of human FFAR3/GPR41.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005295
Test Specificity Clone 16F4.1 targets an epitope in the C-terminal cytoplasmic domain of human FFAR3/GPR41.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.