Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FFN 511, Tocris Bioscience™

Catalog No. 387810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-781-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 38-781-0 Supplier R&D Systems Supplier No. 3878/10
This item is not returnable. View return policy

Fluorescent substrate for VMAT2

Fluorescent false neurotransmitter (FFN). Targets neuronal vesicular monoamine transporter (VMAT) 2; inhibits serotonin binding to VMAT2 (IC50 = 1 μM). Excitation and emission maxima are 406 and 501 nm in pH 7 buffer, respectively.

Chemical Identifiers

CAS 1004548-96-2
Molecular Formula C19H21F3N2O4
Molecular Weight (g/mol) 398.382
InChI Key XHUSAIWAUAMZRG-UHFFFAOYSA-N
Synonym ffn511 tfa salt
PubChem CID 91885442
SMILES C1CC2=C3C(=C4C(=C2)C(=CC(=O)O4)CCN)CCCN3C1.C(=O)(C(F)(F)F)O

Specifications

Quantity 10 mg
Formula Weight 284.35
Percent Purity >99%
Chemical Name or Material FFN 511
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.